Press Releases
RADNOR, Pa. (USA) — July 12, 2021 — Avantor, Inc., a leading global provider of mission critical products and services to customers in the life sciences and advanced technologies & applied materials industries, has introduced J.T.Baker premium robotic tips for use with leading robotic liquid handling and research workstations. Engineered to help scientists and researchers move from discovery to delivery faster, the tips are ideal for a range of applications, from genomics and cell biology to proteomic workflows.
“Our new, high-quality J.T.Baker robotic tips help researchers get the reliable, repeatable results required to keep critical life sciences work moving forward,” said Rohit Shroff, Senior Vice President, Global Lab Products, Avantor. “Our tips provide customers the precision and reliability they need to support their dynamic requirements.”
J.T.Baker robotic tips are designed and manufactured according to rigorous quality standards to help maximize robotic tool utilization and minimize lab downtime.
Avantor leverages decades of engineering and manufacturing expertise from its new Germany-based production team and facilities following the company’s recent acquisition of Ritter GmbH and its affiliates, one of the fastest growing manufacturers of high-quality robotic and liquid handling consumables, including conductive tips engineered to exacting standards. The ISO 9001 and ISO 13485 certified facilities offer a fully automated production line operated under clean room conditions for compatibility and cleanliness. Each batch of J.T.Baker robotic tips are independently tested to be free from DNase, RNase, ATP and pyrogens, to protect against contamination and ensure the highest levels of reliability and repeatability.
The tips are offered in two formats:
- J.T.Baker conductive robotic tips are engineered to minimize downtime and keep operations running smoothly. Their proven design enables even small liquid volumes to be sampled with high precision and without contamination. The Hamilton and Tecan-type tips can be used in many robotic workstations.
- J.T.Baker clear robotic tips provide superior performance for both sample preparation and drug discovery applications. The transparent tips enable visual recognition of foam to help ensure proper sampling.
The tips are part of a full line of automated precision plastics now being added to the company’s globally recognized family of J.T.Baker products. For more than a century, researchers and manufacturers have trusted the application-optimized and function-tested performance of J.T.Baker brand materials to meet the demanding needs of a wide range of applications.
J.T.Baker conductive and clear robotic tips are being supported for easy, convenient global ordering and delivery through Avantor’s market-leading e-commerce channel, vwr.com. For more information, visit vwr.com/precision-plastics.
About Avantor
Avantor®, a Fortune 500 company, is a leading global provider of mission-critical products and services to customers in the biopharma, healthcare, education & government, and advanced technologies & applied materials industries. Our portfolio is used in virtually every stage of the most important research, development and production activities in the industries we serve. Our global footprint enables us to serve more than 225,000 customer locations and gives us extensive access to research laboratories and scientists in more than 180 countries. We set science in motion to create a better world. For more information, please visit www.avantorsciences.com.
Global Lab Products Media Contact
Lynn Homann
Director, Global Marketing Communications
Avantor
M: 215-858-1020
Lynn.Homann@avantorsciences.com
###